Assay Details
| Gene Symbol: RPL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174760.2 | NP_777185.1 | 4 - 5 | 365 | 144 | |
| GenBank mRNA | AB098764.1 | - | 2 - 3 | 328 | 144 | |
| AF143815.1 | - | 4 - 5 | 365 | 144 | ||
| AY911380.1 | - | 5 - 6 | 373 | 144 | ||
| BC104528.1 | - | 1 | 356 | 144 | ||
| BT021081.1 | - | 4 - 5 | 345 | 144 | ||
More Information
Set Membership: |
|

RPL10
Genomic Map