Assay Details
| Gene Symbol: CERT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174765.3 | NP_777190.1 | 9 - 10 | 1437 | 91 | |
| GenBank mRNA | AF232931.1 | - | 9 - 10 | 1438 | 91 | |
| AF232933.1 | - | 9 - 10 | 1438 | 91 | ||
More Information
Set Membership: |
|

CERT1
Genomic Map