Assay Details
| Gene Symbol: RPS27A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174778.1 | NP_777203.1 | 3 - 4 | 190 | 76 | |
| GenBank mRNA | AB098891.1 | - | 3 - 4 | 317 | 76 | |
| AF058700.1 | - | 3 - 4 | 190 | 76 | ||
| BC102491.1 | - | 4 - 5 | 267 | 76 | ||
More Information
Set Membership: |
|

RPS27A
Genomic Map