Assay Details
| Gene Symbol: ATP6V1E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174810.2 | NP_777235.1 | 5 - 6 | 435 | 78 | |
| GenBank mRNA | BC102616.1 | - | 5 - 6 | 435 | 78 | |
| BT021530.1 | - | 5 - 6 | 448 | 78 | ||
| J03244.1 | - | 5 - 6 | 435 | 78 | ||
More Information
Set Membership: |
|

ATP6V1E1
Genomic Map