Assay Details
| Gene Symbol: EIF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174830.1 | NP_777255.1 | 6 - 7 | 752 | 68 | |
| GenBank mRNA | AF141872.1 | - | 6 - 7 | 752 | 68 | |
| BC109922.1 | - | 6 - 7 | 68 | |||
More Information
Set Membership: |
|

EIF6
Genomic Map