Assay Details
| Gene Symbol: NDUFA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175791.2 | NP_786985.1 | 2 - 3 | 259 | 74 | |
| GenBank mRNA | BC102430.1 | - | 2 - 3 | 274 | 74 | |
| X63211.1 | - | 259 | 74 | |||
More Information
Set Membership: |
|

NDUFA6
Genomic Map