Assay Details
| Gene Symbol: RAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175792.2 | NP_786986.1 | 5 - 6 | 562 | 78 | |
| GenBank mRNA | AF175263.1 | - | 5 - 6 | 562 | 78 | |
| BC102255.1 | - | 5 - 6 | 523 | 78 | ||
| BC151444.1 | - | 5 - 6 | 594 | 78 | ||
More Information
Set Membership: |
|

RAC2
Genomic Map