Assay Details
Gene Symbol: NR2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175804.2 | NP_786998.1 | 1 - 2 | 576 | 96 | |
GenBank mRNA | AJ249440.1 | - | 1 - 2 | 576 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
