Assay Details
Gene Symbol: HINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175812.2 | NP_787006.1 | 1 | 107 | 103 | |
GenBank mRNA | BC102553.1 | - | 1 - 2 | 135 | 103 | |
U09405.1 | - | 1 | 107 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
