Assay Details
Gene Symbol: HINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175812.2 | NP_787006.1 | 1 | 219 | 119 | |
GenBank mRNA | BC102553.1 | - | 2 - 3 | 247 | 119 | |
U09405.1 | - | 1 | 219 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
