Assay Details
| Gene Symbol: ATP5ME | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176639.2 | NP_788812.1 | 1 - 2 | 71 | 55 | |
| GenBank mRNA | BC102279.1 | - | 1 - 2 | 70 | 55 | |
| M64751.1 | - | 1 - 2 | 71 | 55 | ||
More Information
Set Membership: |
|

ATP5ME
Genomic Map