Assay Details
| Gene Symbol: FUT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176851.1 | NP_789821.1 | 4 - 5 | 448 | 62 | |
| GenBank mRNA | AJ132773.2 | - | 4 - 5 | 448 | 62 | |
| AJ132774.1 | - | 2 - 3 | 219 | 62 | ||
| AJ132775.1 | - | 2 - 3 | 194 | 62 | ||
| AJ132776.1 | - | 2 - 3 | 188 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ132773.2, AJ132774.1, AJ132775.1, AJ132776.1, NM_176851.1 transcripts.
Set Membership: |
|

FUT6
Genomic Map