Assay Details
| Gene Symbol: TGM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177507.2 | NP_803473.1 | 2 - 3 | 231 | 59 | |
| GenBank mRNA | BC103290.1 | - | 2 - 3 | 240 | 59 | |
| X60686.1 | - | 231 | 59 | |||
More Information
Set Membership: |
|

TGM2
Genomic Map