Assay Details
| Gene Symbol: GLYAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177513.2 | NP_803479.1 | 1 - 2 | 106 | 81 | |
| GenBank mRNA | AF045032.1 | - | 1 - 2 | 106 | 81 | |
| AJ223301.1 | - | 1 - 2 | 106 | 81 | ||
| BC112536.1 | - | 1 - 2 | 126 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ223301.1, BC112536.1, NM_177513.2 transcripts.
Set Membership: |
|

GLYAT
Genomic Map