Assay Details
Gene Symbol: SULT1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177521.2 | NP_803487.1 | 3 - 4 | 409 | 73 | |
GenBank mRNA | BC102274.1 | - | 5 - 6 | 730 | 73 | |
U35253.1 | - | 3 - 4 | 409 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
