Assay Details
| Gene Symbol: AQP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317792.2 | NP_001304721.1 | 4 - 5 | 759 | 72 | |
| NM_001317794.2 | NP_001304723.1 | 4 - 5 | 723 | 72 | ||
| NM_181003.4 | NP_851346.1 | 4 - 5 | 759 | 72 | ||
| GenBank mRNA | AB012950.1 | - | 4 - 5 | 696 | 72 | |
| AB015947.2 | - | 4 - 5 | 762 | 72 | ||
| AB028642.1 | - | 4 - 5 | 703 | 72 | ||
| BC118415.1 | - | 4 - 5 | 759 | 72 | ||
More Information
Set Membership: |
|

AQP4
Genomic Map