Assay Details
Gene Symbol: AKR1C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181027.2 | NP_851370.1 | 8 - 9 | 938 | 103 | |
GenBank mRNA | BC112519.1 | - | 8 - 9 | 963 | 103 | |
D49542.1 | - | 8 - 9 | 938 | 103 | ||
D88749.1 | - | 8 - 9 | 939 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
