Assay Details
Gene Symbol: GAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203358.2 | NP_976234.2 | 2 - 3 | 791 | 92 | |
GenBank mRNA | BC118452.1 | - | 2 - 3 | 700 | 92 | |
DV906486.1 | - | 2 - 3 | 748 | 92 | ||
DV912723.1 | - | 1 - 2 | 646 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
