Assay Details
| Gene Symbol: SLC5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203360.1 | NP_976236.1 | 12 - 13 | 1678 | 58 | |
| GenBank mRNA | AY208941.1 | - | 12 - 13 | 1678 | 58 | |
More Information
Set Membership: |
|

SLC5A2
Genomic Map