Assay Details
| Gene Symbol: RPL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205797.1 | NP_991366.1 | 1 | 164 | 129 | |
| GenBank mRNA | AY528249.1 | - | 1 | 164 | 129 | |
| BC102693.1 | - | 1 | 197 | 129 | ||
| BC142032.1 | - | 1 | 197 | 129 | ||
| Gene Symbol: LOC784762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB098966.1 | - | 1 | 164 | 129 | |
More Information
Set Membership: |
|

RPL12
Genomic Map