Assay Details
| Gene Symbol: NME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205818.2 | NP_991387.2 | 1 - 2 | 111 | 57 | |
| GenBank mRNA | CR454063.1 | - | 1 - 2 | 92 | 57 | |
| EE337437.1 | - | 1 - 2 | 112 | 57 | ||
| X92956.1 | - | 50 | 57 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_205818.2, X92956.1 transcripts.
Set Membership: |
|

NME1
Genomic Map