Assay Details
| Gene Symbol: HSD11B1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206969.1 | NP_996852.1 | 3 - 4 | 327 | 83 | |
| GenBank mRNA | AY554049.1 | - | 3 - 4 | 327 | 83 | |
More Information
Set Membership: |
|

HSD11B1L
Genomic Map