Assay Details
Gene Symbol: KLRJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002884.1 | NP_001002884.1 | 7 - 8 | 778 | 93 | |
GenBank mRNA | AY156928.1 | - | 7 - 8 | 778 | 93 | |
BC149263.1 | - | 7 - 8 | 844 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
