Assay Details
| Gene Symbol: KLRJ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002884.1 | NP_001002884.1 | 8 - 9 | 934 | 136 | |
| GenBank mRNA | AY156928.1 | - | 8 - 9 | 934 | 136 | |
| BC149263.1 | - | 8 - 9 | 1000 | 136 | ||
More Information
Set Membership: |
|

KLRJ1
Genomic Map