Assay Details
Gene Symbol: KLRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002890.1 | NP_001002890.1 | 3 - 4 | 208 | 117 | |
GenBank mRNA | AF422180.1 | - | 3 - 4 | 208 | 117 | |
EF081290.1 | - | 2 - 3 | 153 | 117 | ||
EF081291.1 | - | 2 - 3 | 153 | 117 | ||
EF081292.1 | - | 2 - 3 | 153 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
