Assay Details
Gene Symbol: CST6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012764.3 | NP_001012782.1 | 2 - 3 | 406 | 60 | |
GenBank mRNA | EE220848.1 | - | 2 - 3 | 407 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
