Assay Details
| Gene Symbol: DXO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014872.2 | NP_001014872.2 | 9 - 10 | 2140 | 76 | |
| GenBank mRNA | BT021779.1 | - | 9 - 10 | 2140 | 76 | |
More Information
Set Membership: |
|

DXO
Genomic Map