Assay Details
Gene Symbol: MTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014874.1 | NP_001014874.1 | 2 - 3 | 267 | 59 | |
GenBank mRNA | BT021695.1 | - | 2 - 3 | 267 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
