Assay Details
| Gene Symbol: NR1H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014883.1 | NP_001014883.1 | 4 - 5 | 431 | 72 | |
| GenBank mRNA | BC109925.1 | - | 4 - 5 | 427 | 72 | |
| BT021148.1 | - | 4 - 5 | 419 | 72 | ||
| BT021903.1 | - | 4 - 5 | 431 | 72 | ||
More Information
Set Membership: |
|

NR1H2
Genomic Map