Assay Details
Gene Symbol: PRICKLE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014886.1 | NP_001014886.1 | 2 - 3 | 195 | 61 | |
GenBank mRNA | BT021566.1 | - | 2 - 3 | 195 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
