Assay Details
| Gene Symbol: ALAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014895.1 | NP_001014895.1 | 2 - 3 | 250 | 66 | |
| GenBank mRNA | BC112596.1 | - | 2 - 3 | 216 | 66 | |
| BT021592.1 | - | 2 - 3 | 250 | 66 | ||
More Information
Set Membership: |
|

ALAD
Genomic Map