Assay Details
| Gene Symbol: HDHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014896.1 | NP_001014896.1 | 1 - 2 | 147 | 68 | |
| GenBank mRNA | BC102640.1 | - | 1 - 2 | 136 | 68 | |
| BT020984.1 | - | 1 - 2 | 147 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102640.1, NM_001014896.1 transcripts.
Set Membership: |
|

HDHD3
Genomic Map