Assay Details
Gene Symbol: TCF21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014899.1 | NP_001014899.1 | 1 - 2 | 701 | 85 | |
GenBank mRNA | BC105574.1 | - | 1 - 2 | 706 | 85 | |
BT020847.1 | - | 1 - 2 | 701 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
