Assay Details
| Gene Symbol: PYCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014906.1 | NP_001014906.1 | 5 - 6 | 657 | 69 | |
| GenBank mRNA | BC149378.1 | - | 5 - 6 | 670 | 69 | |
| BT021789.1 | - | 5 - 6 | 657 | 69 | ||
More Information
Set Membership: |
|

PYCR3
Genomic Map