Assay Details
| Gene Symbol: HMOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014912.1 | NP_001014912.1 | 1 - 2 | 121 | 81 | |
| GenBank mRNA | BC102105.1 | - | 1 - 2 | 139 | 81 | |
| BT020968.1 | - | 1 - 2 | 121 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102105.1, NM_001014912.1 transcripts.
Set Membership: |
|

HMOX1
Genomic Map