Assay Details
| Gene Symbol: NFE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014923.1 | NP_001014923.1 | 1 - 2 | 37 | 57 | |
| GenBank mRNA | BT021174.1 | - | 1 - 2 | 37 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001014923.1 transcripts.
Set Membership: |
|

NFE2
Genomic Map