Assay Details
| Gene Symbol: NFE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014923.1 | NP_001014923.1 | 2 - 3 | 201 | 63 | |
| GenBank mRNA | BT020636.1 | - | 2 - 3 | 205 | 63 | |
| BT021174.1 | - | 2 - 3 | 201 | 63 | ||
More Information
Set Membership: |
|

NFE2
Genomic Map