Assay Details
| Gene Symbol: EEF1B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014936.1 | NP_001014936.1 | 3 - 4 | 296 | 82 | |
| GenBank mRNA | AY911340.1 | - | 2 - 3 | 292 | 82 | |
| BC102548.1 | - | 2 - 3 | 217 | 82 | ||
| BC147944.1 | - | 2 - 3 | 332 | 82 | ||
| BT021037.1 | - | 3 - 4 | 296 | 82 | ||
More Information
Set Membership: |
|

EEF1B2
Genomic Map