Assay Details
Gene Symbol: EEF1B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014936.1 | NP_001014936.1 | 6 - 7 | 618 | 67 | |
GenBank mRNA | AY911340.1 | - | 5 - 6 | 614 | 67 | |
BC102548.1 | - | 5 - 6 | 539 | 67 | ||
BC147944.1 | - | 5 - 6 | 654 | 67 | ||
BT021037.1 | - | 6 - 7 | 618 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
