Assay Details
| Gene Symbol: RPS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015548.2 | NP_001015548.1 | 4 - 5 | 583 | 64 | |
| GenBank mRNA | AY911358.1 | - | 4 - 5 | 538 | 64 | |
| BC102493.1 | - | 1 | 538 | 64 | ||
| BT021025.1 | - | 4 - 5 | 525 | 64 | ||
| EE223032.1 | - | 4 - 5 | 584 | 64 | ||
More Information
Set Membership: |
|

RPS6
Genomic Map