Assay Details
| Gene Symbol: HNF4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015557.1 | NP_001015557.1 | 6 - 7 | 728 | 60 | |
| GenBank mRNA | AF250028.1 | - | 2 - 3 | 164 | 60 | |
| AY318752.1 | - | 6 - 7 | 728 | 60 | ||
| DQ192586.1 | - | 4 - 5 | 409 | 60 | ||
More Information
Set Membership: |
|

HNF4A
Genomic Map