Assay Details
| Gene Symbol: BRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015582.1 | NP_001015582.1 | 3 - 4 | 586 | 78 | |
| GenBank mRNA | BC113270.1 | - | 3 - 4 | 565 | 78 | |
| BT020651.1 | - | 3 - 4 | 578 | 78 | ||
| BT020781.1 | - | 3 - 4 | 569 | 78 | ||
| BT020876.1 | - | 3 - 4 | 586 | 78 | ||
| BT020884.1 | - | 3 - 4 | 569 | 78 | ||
| BT020898.1 | - | 3 - 4 | 570 | 78 | ||
More Information
Set Membership: |
|

BRF2
Genomic Map