Assay Details
| Gene Symbol: DDA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015583.1 | NP_001015583.1 | 2 - 3 | 204 | 68 | |
| GenBank mRNA | BC120366.1 | - | 2 - 3 | 145 | 68 | |
| BT021011.1 | - | 2 - 3 | 204 | 68 | ||
More Information
Set Membership: |
|

DDA1
Genomic Map