Assay Details
| Gene Symbol: GDPGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015629.1 | NP_001015629.1 | 2 - 3 | 407 | 94 | |
| GenBank mRNA | BT020839.1 | - | 2 - 3 | 407 | 94 | |
| BT021157.1 | - | 1 - 2 | 104 | 94 | ||
| BT021160.1 | - | 2 - 3 | 405 | 94 | ||
| BT021723.1 | - | 2 - 3 | 455 | 94 | ||
| BT026292.1 | - | 2 - 3 | 371 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001015629.1 transcripts.
Set Membership: |
|

GDPGP1
Genomic Map