Assay Details
| Gene Symbol: RPL10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015647.1 | NP_001015647.1 | 5 - 6 | 500 | 67 | |
| GenBank mRNA | BC102653.1 | - | 5 - 6 | 515 | 67 | |
| BT020974.1 | - | 5 - 6 | 500 | 67 | ||
More Information
Set Membership: |
|

RPL10A
Genomic Map