Assay Details
| Gene Symbol: UFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015663.1 | NP_001015663.1 | 1 - 2 | 133 | 74 | |
| GenBank mRNA | BC103142.1 | - | 1 - 2 | 141 | 74 | |
| BT021067.1 | - | 1 - 2 | 133 | 74 | ||
More Information
Set Membership: |
|

UFC1
Genomic Map