Assay Details
Gene Symbol: C3H1orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015666.1 | NP_001015666.1 | 6 - 7 | 720 | 78 | |
GenBank mRNA | BC118321.1 | - | 6 - 7 | 707 | 78 | |
BT021077.1 | - | 6 - 7 | 720 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
