Assay Details
| Gene Symbol: DTX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017945.1 | NP_001017945.1 | 8 - 9 | 1622 | 67 | |
| GenBank mRNA | BC103233.1 | - | 7 - 8 | 1041 | 67 | |
| BT021146.1 | - | 8 - 9 | 1622 | 67 | ||
More Information
Set Membership: |
|

DTX2
Genomic Map