Assay Details
| Gene Symbol: IQCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024492.1 | NP_001019663.1 | 2 - 3 | 977 | 69 | |
| GenBank mRNA | BT021671.1 | - | 2 - 3 | 977 | 69 | |
More Information
Set Membership: |
|

IQCC
Genomic Map