Assay Details
| Gene Symbol: MRPS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024500.2 | NP_001019671.1 | 1 - 2 | 109 | 99 | |
| GenBank mRNA | AY911326.1 | - | 1 - 2 | 110 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001024500.2 transcripts.
Set Membership: |
|

MRPS17
Genomic Map